Gene Transcripts
- Gene Symbol
- Nmral1
- Entrez Gene ID
- 67824
- Gene Name
- NmrA-like family domain containing 1
- Gene Aliases
- 1110025F24Rik, AI256624
- Chromosome Location
- Chr. 16: 4711317 - 4719356 on Build GRCm38
- UniGene ID
- Mm.28451
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026393.2 |
NP_080669.1
|
3, 4 |
740 |
|
GenBank mRNA |
BC030039.1 |
- |
3, 4 |
443 |
|
View Full Product Details