Gene Transcripts
- Gene Symbol
- PEX7
- Entrez Gene ID
- 5191
- Gene Name
- peroxisomal biogenesis factor 7
- Gene Aliases
- PBD9B, PTS2R, RCDP1, RD
- Chromosome Location
- Chr. 6: 136822220 - 136913934 on Build GRCh38
- UniGene ID
- Hs.280932
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000288.3 |
NP_000279.1
|
2, 3 |
286 |
|
XM_005267019.4 |
XP_005267076.1
|
2, 3 |
566 |
|
XM_006715502.1 |
XP_006715565.1
|
2, 3 |
630 |
|
XM_017010934.1 |
XP_016866423.1
|
2, 3 |
630 |
|
GenBank mRNA |
BC006268.1 |
- |
2, 3 |
228 |
|
BC031606.1 |
- |
2, 3 |
270 |
5176358 |
BP246238.1 |
- |
2, 3 |
286 |
|
CR542054.1 |
- |
2, 3 |
184 |
|
CR542076.1 |
- |
2, 3 |
184 |
|
U76560.1 |
- |
2, 3 |
232 |
|
U88871.1 |
- |
2, 3 |
265 |
|
View Full Product Details