Gene Transcripts

Gene Symbol
Spsb3
Entrez Gene ID
79043
Gene Name
splA/ryanodine receptor domain and SOCS box containing 3
Gene Aliases
2310012N15Rik, 3300001M01Rik, SSB3, Tce1, mKIAA4204
Chromosome Location
Chr. 17: 24886349 - 24892147 on Build GRCm38
UniGene ID
Mm.274794
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163750.1 NP_001157222.1 3, 4 746
NM_001163751.1 NP_001157223.1 3, 4 401
NM_027141.2 NP_081417.2 3, 4 422
GenBank mRNA AF403038.1 - 3, 4 422
AK009325.1 - 3, 4 422
AK076170.1 - 3, 4 698
AK082768.1 - 3, 4 345
AK167474.1 - 3, 4 662
AK184627.1 - 1, 2 20
AK220234.1 - 3, 4 684
BC072556.1 - 3, 4 707 5691114
BY276751.1 - 3, 4 402

View Full Product Details