Gene Transcripts
- Gene Symbol
- Parp8
- Entrez Gene ID
- 52552
- Gene Name
- poly (ADP-ribose) polymerase family, member 8
- Gene Aliases
- 2810430O08Rik, ARTD16, D13Ertd275e
- Chromosome Location
- Chr. 13: 116850842 - 117025913 on Build GRCm38
- UniGene ID
- Mm.274466
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001081009.1 |
NP_001074478.1
|
16, 17 |
1833 |
|
GenBank mRNA |
AK149889.1 |
- |
15, 16 |
1800 |
|
AK149918.1 |
- |
15, 16 |
1800 |
|
AK150208.1 |
- |
15, 16 |
1800 |
|
AK150732.1 |
- |
14, 15 |
1610 |
|
AK151510.1 |
- |
14, 15 |
1611 |
|
AK152919.1 |
- |
14, 15 |
1610 |
|
AK153169.1 |
- |
14, 15 |
1610 |
|
AK153270.1 |
- |
14, 15 |
1610 |
|
BC021315.1 |
- |
6, 7 |
967 |
5027417 |
BC021881.1 |
- |
15, 16 |
1803 |
3710450 |
BC022679.1 |
- |
1, 2 |
360 |
|
EF591877.1 |
- |
|
1568 |
|
View Full Product Details