Gene Transcripts

Gene Symbol
PDHA1
Entrez Gene ID
5160
Gene Name
pyruvate dehydrogenase (lipoamide) alpha 1
Gene Aliases
PDHA, PDHAD, PDHCE1A, PHE1A
Chromosome Location
Chr. X: 19343893 - 19361707 on Build GRCh38
UniGene ID
Hs.530331
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000284.3 NP_000275.1 3 330
NM_001173454.1 NP_001166925.1 4 444
NM_001173455.1 NP_001166926.1 3 330
NM_001173456.1 NP_001166927.1 3 330
XM_017029574.1 XP_016885063.1 4 431
GenBank mRNA AK129887.1 - 3 265
AK222740.1 - 3 261
AK222988.1 - 3 295
AK293250.1 - 4 409
AK296341.1 - 3 296
AK296457.1 - 3 244
AK312263.1 - 3 296
BC002406.1 - 3 242 2961286
DC361191.1 - 3 330
EF590117.1 - 362
J03503.1 - 3 257
J03575.1 - 3 230
L13318.1 - 3 290
L48690.1 - 3 290
M24848.1 - 3 228
X52709.1 - 3 239
X52710.1 - 3 241

View Full Product Details