Gene Transcripts

Gene Symbol
Spats1
Entrez Gene ID
71020
Gene Name
spermatogenesis associated, serine-rich 1
Gene Aliases
1700011H05Rik, 4933400B06Rik, Daip2, Ddip, Srsp1
Chromosome Location
Chr. 17: 45445877 - 45475194 on Build GRCm38
UniGene ID
Mm.44257
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027649.3 NP_081925.2 3 408
GenBank mRNA AK005865.1 - 3 409
AK016586.1 - 3 335
AK078278.1 - 330
AK165435.1 - 2 196
AY336501.1 - 3 296

View Full Product Details