Gene Transcripts

Gene Symbol
Sntg1
Entrez Gene ID
71096
Gene Name
syntrophin, gamma 1
Gene Aliases
4933426D16Rik, G1SYN, SYN4
Chromosome Location
Chr. 1: 8359739 - 9301061 on Build GRCm38
UniGene ID
Mm.336395
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290390.1 NP_001277319.1 7 2179
NM_001290392.1 NP_001277321.1 6 2125
NM_001290393.1 NP_001277322.1 6 2125
NM_027671.5 NP_081947.2 7 2179
GenBank mRNA AF367759.1 - 6 831
AK038698.1 - 6 2126
AK045676.1 - 6 1510
AK080034.1 - 7 1541
AK083241.1 - 7 1167

View Full Product Details