Gene Transcripts
- Gene Symbol
- March8
- Entrez Gene ID
- 71779
- Gene Name
- membrane-associated ring finger (C3HC4) 8
- Gene Aliases
- 1300017E09Rik, MARCH-VIII, Mir
- Chromosome Location
- Chr. 6: 116337884 - 116409540 on Build GRCm38
- UniGene ID
- Mm.27064
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK005032.2 |
- |
5, 6 |
601 |
|
AK154976.1 |
- |
6, 7 |
793 |
|
AK165647.1 |
- |
5, 6 |
590 |
|
AK182250.1 |
- |
4, 5 |
374 |
|
AK189694.1 |
- |
4, 5 |
376 |
|
AK197895.1 |
- |
3, 4 |
291 |
|
AK209829.1 |
- |
2, 3 |
280 |
|
AK210194.1 |
- |
3, 4 |
290 |
|
BC050908.1 |
- |
6, 7 |
776 |
6416955 |
BC053090.1 |
- |
5, 6 |
861 |
5709246 |
BQ770041.1 |
- |
5, 6 |
660 |
|
View Full Product Details