Gene Transcripts

Gene Symbol
Frmd6
Entrez Gene ID
319710
Gene Name
FERM domain containing 6
Gene Aliases
2610019M19Rik, 4930488L10Rik, AW212977
Chromosome Location
Chr. 12: 70825335 - 70902234 on Build GRCm38
UniGene ID
Mm.2962
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028127.3 NP_082403.2 14 4238
GenBank mRNA AK011469.1 - 1 1602
BC019939.1 - 1 1883
BC053929.1 - 14 4238 6389695

View Full Product Details