Gene Transcripts

Gene Symbol
Efhd1
Entrez Gene ID
98363
Gene Name
EF hand domain containing 1
Gene Aliases
4931430I01Rik, AI452351, PP3051, mitocalcin
Chromosome Location
Chr. 1: 87264308 - 87310839 on Build GRCm38
UniGene ID
Mm.247951
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028889.2 NP_083165.1 1 325
GenBank mRNA AK016491.1 - 1 325
AK045911.1 - 1 327
AK144175.1 - 1 328
AK171846.1 - 1 327
BC019531.1 - 1 311 4459208
BC085088.1 - 1 310 6467625

View Full Product Details