Gene Transcripts

Gene Symbol
Armc8
Entrez Gene ID
74125
Gene Name
armadillo repeat containing 8
Gene Aliases
1200015K23Rik, Gid5, HSPC056
Chromosome Location
Chr. 9: 99478385 - 99568936 on Build GRCm38
UniGene ID
Mm.234823
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166138.1 NP_001159610.1 11, 12 1298
NM_028768.3 NP_083044.2 11, 12 1298
GenBank mRNA AK004793.2 - 11, 12 1299
AK078280.1 - 11, 12 1049
AK083094.1 - 10, 11 990
BC038866.1 - 11, 12 1256 1230321

View Full Product Details