Gene Transcripts

Gene Symbol
Prkd3
Entrez Gene ID
75292
Gene Name
protein kinase D3
Gene Aliases
4930557O20Rik, 5730497N19Rik, PKD3, Pkcnu, Prkcn
Chromosome Location
Chr. 17: 78949405 - 79020816 on Build GRCm38
UniGene ID
Mm.252776
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171004.1 NP_001164475.1 2 933
NM_029239.3 NP_083515.2 2 933
GenBank mRNA AK138826.1 - 2 898
BC027320.1 - 1 334
BC037012.1 - 1 577 5342428
BC089164.1 - 2 832 5400250

View Full Product Details