Gene Transcripts

Gene Symbol
Prkd3
Entrez Gene ID
75292
Gene Name
protein kinase D3
Gene Aliases
4930557O20Rik, 5730497N19Rik, PKD3, Pkcnu, Prkcn
Chromosome Location
Chr. 17: 78949405 - 79020816 on Build GRCm38
UniGene ID
Mm.252776
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171004.1 NP_001164475.1 4, 5 1314
NM_001171005.1 NP_001164476.1 3, 4 510
NM_029239.3 NP_083515.2 4, 5 1314
GenBank mRNA AK016167.1 - 4, 5 469
AK030058.1 - 3, 4 507
AK050059.1 - 2, 3 338
BC027320.1 - 3, 4 713
BC037012.1 - 3, 4 958 5342428
BC089164.1 - 4, 5 1213 5400250

View Full Product Details