Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256100.1 | NP_001243029.1 | 3 | 438 | |
NM_001256101.1 | NP_001243030.1 | 3 | 427 | ||
NM_001256102.1 | NP_001243031.1 | 2 | 267 | ||
NM_029581.4 | NP_083857.1 | 2 | 278 | ||
GenBank mRNA | AK012733.1 | - | 3 | 405 | |
AK147090.1 | - | 2 | 256 | ||
BC052439.1 | - | 2 | 232 | 6406303 | |
BC115684.1 | - | 2 | 219 | 40104230 | |
BC115685.1 | - | 3 | 390 | 40104232 | |
BY284518.1 | - | 2 | 267 |