Gene Transcripts

Gene Symbol
Nudt16
Entrez Gene ID
75686
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 16
Gene Aliases
2310041H06Rik, 2810047L04Rik, 2900006H04Rik, AI851783
Chromosome Location
Chr. 9: 105129338 - 105131841 on Build GRCm38
UniGene ID
Mm.305810
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029385.2 NP_083661.2 3 740
GenBank mRNA AK009737.1 - 3 746
AK012920.1 - 2 793
AK013493.1 - 1 1576
BC064048.1 - 3 727 574013

View Full Product Details