Gene Transcripts

Gene Symbol
Plekha2
Entrez Gene ID
83436
Gene Name
pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 2
Gene Aliases
6430512N22Rik, AI851530, AU041791, TAPP2, W91773
Chromosome Location
Chr. 8: 25039144 - 25104917 on Build GRCm38
UniGene ID
Mm.261122
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031257.3 NP_112547.1 12 1954
GenBank mRNA AF418551.1 - 11 1819
AK032325.1 - 12 1963
AK078212.1 - 12 1933
AK088395.1 - 12 1896
AK153117.1 - 12 1926
BC010215.1 - 12 1875 3498236

View Full Product Details