Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032544.3 | NP_115933.2 | 3 | 1017 | |
GenBank mRNA | AF361482.2 | - | 3 | 1017 | |
AK046330.1 | - | 3 | 393 | ||
BC055716.1 | - | 3 | 369 | ||
BC075712.1 | - | 3 | 377 | ||
BC092042.1 | - | 3 | 400 | ||
BC138560.1 | - | 3 | 426 | 8861580 | |
BC145333.1 | - | 3 | 426 |