Gene Transcripts

Gene Symbol
Myb
Entrez Gene ID
17863
Gene Name
myeloblastosis oncogene
Gene Aliases
AI550390, M16449, c-myb
Chromosome Location
Chr. 10: 21124930 - 21160984 on Build GRCm38
UniGene ID
Mm.473872
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198914.1 NP_001185843.1 16 2985
NM_010848.3 NP_034978.3 15 2628
GenBank mRNA AK088020.1 - 2 655
AK088079.1 - 16 2905
AK132250.1 - 15 2493
AK137794.1 - 9 1574
M12848.1 - 15 2626
X02774.1 - 15 2397

View Full Product Details