Gene Transcripts
- Gene Symbol
- Lyrm5
- Entrez Gene ID
- 67636
- Gene Name
- LYR motif containing 5
- Gene Aliases
- 4930469P12Rik, AL022899, Ghiso
- Chromosome Location
- Chr. 6: 145211134 - 145216542 on Build GRCm38
- UniGene ID
- Mm.486359
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF412298.1 |
- |
3 |
2054 |
|
View Full Product Details