Gene Transcripts

Gene Symbol
Metrnl
Entrez Gene ID
210029
Gene Name
meteorin, glial cell differentiation regulator-like
Gene Aliases
9430048M07Rik, BC019776
Chromosome Location
Chr. 11: 121702427 - 121717389 on Build GRCm38
UniGene ID
Mm.153566
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144797.3 NP_659046.1 4 1894
GenBank mRNA AK034855.1 - 4 1895

View Full Product Details