Gene Transcripts

Gene Symbol
Nrbp2
Entrez Gene ID
223649
Gene Name
nuclear receptor binding protein 2
Gene Aliases
BC011468
Chromosome Location
Chr. 15: 76085589 - 76091594 on Build GRCm38
UniGene ID
Mm.101946
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144847.1 NP_659096.1 6 515
GenBank mRNA BC011468.1 - 6 448
BC012437.1 - 6 515 3710089
BC068117.1 - 11 1163

View Full Product Details