Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001177810.1 | NP_001171281.1 | 5, 6 | 760 | |
NM_001177811.1 | NP_001171282.1 | 5, 6 | 760 | ||
NM_144850.2 | NP_659099.2 | 5, 6 | 760 | ||
GenBank mRNA | AK044566.1 | - | 5, 6 | 683 | |
AK142594.1 | - | 5, 6 | 654 | ||
AK169430.1 | - | 5, 6 | 759 | ||
BC020532.1 | - | 5, 6 | 772 | 4236136 |