Gene Transcripts

Gene Symbol
Ggact
Entrez Gene ID
223267
Gene Name
gamma-glutamylamine cyclotransferase
Gene Aliases
A2ld1
Chromosome Location
Chr. 14: 122890860 - 122913165 on Build GRCm38
UniGene ID
Mm.379358
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145466.2 NP_663441.1 3 620
GenBank mRNA AK050251.1 - 3 610
BB667658.1 - 1 219
BC006662.1 - 3 590 3501534

View Full Product Details