Gene Transcripts

Gene Symbol
Slc35c1
Entrez Gene ID
228368
Gene Name
solute carrier family 35, member C1
Gene Aliases
E430007K15Rik, fuct1
Chromosome Location
Chr. 2: 92452764 - 92460518 on Build GRCm38
UniGene ID
Mm.273990
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145832.3 NP_665831.1 3 678
NM_211358.2 NP_997597.1 2 1166
GenBank mRNA AJ440720.1 - 2 563
AK030977.1 - 3 679
AK033295.1 - 2 255
AK040986.1 - 2 913
AK045972.1 - 3 655
AK050311.1 - 3 643
AK081068.1 - 2 664
AK088227.1 - 3 665
AK138595.1 - 2 1109
AK144762.1 - 2 1088
AK151154.1 - 2 1162
BC031814.1 - 2 760 4196818
BC116736.1 - 2 591 40126055
BC119128.1 - 2 591 8733877

View Full Product Details