Gene Transcripts

Gene Symbol
Krt79
Entrez Gene ID
223917
Gene Name
keratin 79
Gene Aliases
BC031593
Chromosome Location
Chr. 15: 101929332 - 101940324 on Build GRCm38
UniGene ID
Mm.162744
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146063.1 NP_666175.1 9 1844
GenBank mRNA AK076407.1 - 5 900
BC019155.1 - 9 1783 5065687
BC031593.1 - 9 1844 4935395

View Full Product Details