Gene Transcripts

Gene Symbol
RPS16
Entrez Gene ID
6217
Gene Name
ribosomal protein S16
Gene Aliases
S16
Chromosome Location
Chr. 19: 39433137 - 39436020 on Build GRCh38
UniGene ID
Hs.397609
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001020.5 NP_001011.1 3 313
NM_001321111.1 NP_001308040.1 3 262
XM_005259137.2 XP_005259194.1 3 313
GenBank mRNA AK055869.1 - 2 2086
AK315498.1 - 3 241
BC004324.2 - 3 239 3628799
BC007977.2 - 3 224 4310307
BC024970.1 - 1 209
BC071674.1 - 3 242 5394209
BC095429.1 - 3 183
BQ421722.1 - 3 271
CB107158.1 - 3 291
M60854.1 - 3 225

View Full Product Details