Gene Transcripts

Gene Symbol
Mrgpra3
Entrez Gene ID
233222
Gene Name
MAS-related GPR, member A3
Gene Aliases
Mrga3
Chromosome Location
Chr. 7: 47588950 - 47601372 on Build GRCm38
UniGene ID
Mm.240142
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153067.2 NP_694707.2 2 432
GenBank mRNA AY042193.1 - 2 447

View Full Product Details