Gene Transcripts

Gene Symbol
Thsd7b
Entrez Gene ID
210417
Gene Name
thrombospondin, type I, domain containing 7B
Gene Aliases
1700074E13Rik, B930082A18, D130067I03Rik
Chromosome Location
Chr. 1: 129273304 - 130219278 on Build GRCm38
UniGene ID
Mm.380518
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172485.3 NP_766073.2 3 1140
GenBank mRNA AK047508.1 - 926
AK051714.1 - 3 982
AK083798.1 - 3 968
BC063250.1 - 3 970 6410178

View Full Product Details