Gene Transcripts

Gene Symbol
Rab39b
Entrez Gene ID
67790
Gene Name
RAB39B, member RAS oncogene family
Gene Aliases
6330580M05Rik
Chromosome Location
Chr. X: 75572045 - 75578231 on Build GRCm38
UniGene ID
Mm.45148
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175122.6 NP_780331.1 1 395
GenBank mRNA AB232636.1 - 1 171
AK018257.1 - 1 380
AK020665.1 - 1 376
AK039013.1 - 1 369
AK048981.1 - 1 369
AK160737.1 - 1 369
BC050853.1 - 1 389 6517891
BC052472.1 - 1 320 6833461
BY257784.1 - 1 395

View Full Product Details