Gene Transcripts

Gene Symbol
Pou6f2
Entrez Gene ID
218030
Gene Name
POU domain, class 6, transcription factor 2
Gene Aliases
D130006K24Rik, RPF-1
Chromosome Location
Chr. 13: 18124960 - 18382044 on Build GRCm38
UniGene ID
Mm.352263
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175006.2 NP_778171.2 211
GenBank mRNA AK083772.1 - 211
BC094915.1 - 226 6435959
BC138151.1 - 130 8861170

View Full Product Details