Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001098.2 | NP_001089.1 | 4 | 474 | |
XM_017028812.1 | XP_016884301.1 | 4 | 788 | ||
GenBank mRNA | AK296260.1 | - | 5 | 439 | |
AK297213.1 | - | 4 | 349 | ||
AK302691.1 | - | 5 | 424 | ||
AK314845.1 | - | 4 | 482 | ||
BC014092.2 | - | 4 | 474 | 4328775 | |
BC026196.2 | - | 4 | 477 | 5264179 | |
CR456365.1 | - | 4 | 466 | ||
CR536568.1 | - | 4 | 452 | ||
GQ472196.1 | - | 4 | 477 | ||
U80040.1 | - | 4 | 472 |