Gene Transcripts

Gene Symbol
Rnf38
Entrez Gene ID
73469
Gene Name
ring finger protein 38
Gene Aliases
1700065B19Rik, 2610202O07Rik, AA673263, Oip1
Chromosome Location
Chr. 4: 44126212 - 44233643 on Build GRCm38
UniGene ID
Mm.262859
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038993.3 NP_001034082.1 2 111
GenBank mRNA AK147608.1 - 2 168

View Full Product Details