Gene Transcripts

Gene Symbol
Psmb11
Entrez Gene ID
73902
Gene Name
proteasome (prosome, macropain) subunit, beta type, 11
Gene Aliases
5830406J20Rik, beta5t
Chromosome Location
Chr. 14: 54625310 - 54629556 on Build GRCm38
UniGene ID
Mm.32009
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175204.4 NP_780413.1 1 3602
GenBank mRNA AK037368.1 - 1 3599
AK162727.1 - 1 3624

View Full Product Details