Gene Transcripts

Gene Symbol
Txnl4b
Entrez Gene ID
234723
Gene Name
thioredoxin-like 4B
Gene Aliases
AI595343, D530025J19Rik, DLP, Dim2
Chromosome Location
Chr. 8: 109565986 - 109574051 on Build GRCm38
UniGene ID
Mm.37667
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175646.3 NP_783577.2 4 1025
GenBank mRNA AK079533.1 - 4 1124
AK085226.1 - 4 1006
BC094379.1 - 4 1041 30541506
BC138333.1 - 3 950 8861353

View Full Product Details