Gene Transcripts
- Gene Symbol
- Dpep2
- Entrez Gene ID
- 319446
- Gene Name
- dipeptidase 2
- Gene Aliases
- F630103D06Rik, MBD-2
- Chromosome Location
- Chr. 8: 105984945 - 106006348 on Build GRCm38
- UniGene ID
- Mm.248357
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301204.1 |
NP_001288133.1
|
6, 7 |
921 |
|
NM_001301205.1 |
NP_001288134.1
|
5, 6 |
606 |
|
NM_176913.4 |
NP_795887.2
|
6, 7 |
1045 |
|
GenBank mRNA |
AF488552.1 |
- |
6, 7 |
1045 |
|
AK089165.1 |
- |
3, 4 |
2659 |
|
AK089244.1 |
- |
6, 7 |
907 |
|
AK155099.1 |
- |
6, 7 |
873 |
|
AK155308.1 |
- |
6, 7 |
872 |
|
AK155417.1 |
- |
6, 7 |
876 |
|
AK170094.1 |
- |
6, 7 |
873 |
|
AK170970.1 |
- |
6, 7 |
873 |
|
AK171044.1 |
- |
6, 7 |
873 |
|
AK171122.1 |
- |
6, 7 |
873 |
|
AK171155.1 |
- |
5, 6 |
586 |
|
AK171196.1 |
- |
6, 7 |
873 |
|
AK171268.1 |
- |
6, 7 |
873 |
|
AK171503.1 |
- |
6, 7 |
873 |
|
AK171606.1 |
- |
6, 7 |
889 |
|
View Full Product Details