Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177139.5 | NP_796113.1 | 4, 5 | 575 | |
GenBank mRNA | AK046251.1 | - | 1, 2 | 131 | |
AK053620.1 | - | 5, 6 | 706 | ||
BB619054.1 | - | 542 | |||
BB653059.1 | - | 536 | |||
BC070462.1 | - | 4, 5 | 596 | 30650880 |