Gene Transcripts

Gene Symbol
CLNS1A
Entrez Gene ID
1207
Gene Name
chloride nucleotide-sensitive channel 1A
Gene Aliases
CLCI, CLNS1B, ICln
Chromosome Location
Chr. 11: 77616151 - 77637806 on Build GRCh38
UniGene ID
Hs.430733
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293.2 NP_001284.1 4 520
NM_001311199.1 NP_001298128.1 4 520
NM_001311200.1 NP_001298129.1 3 418
NM_001311201.1 NP_001298130.1 4 520
GenBank mRNA AF005422.1 - 4 458
AF026003.1 - 4 428
AK315259.1 - 4 514
AL530926.3 - 3 364
BC119634.1 - 4 477 40115653
BC119635.1 - 4 477 40115655
BQ643786.1 - 4 490
BT019907.1 - 4 428
BU538016.1 - 4 486
U17899.1 - 4 502
U53454.1 - 4 512
X91788.1 - 4 516

View Full Product Details