Gene Transcripts

Gene Symbol
Rinl
Entrez Gene ID
320435
Gene Name
Ras and Rab interactor-like
Gene Aliases
5830482F20Rik, 9930116N10Rik
Chromosome Location
Chr. 7: 28788861 - 28798965 on Build GRCm38
UniGene ID
Mm.74632
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177158.5 NP_796132.2 4 355
GenBank mRNA AK030993.1 - 4 348
BC044761.1 - 4 391
BC046530.1 - 4 391 5329601

View Full Product Details