Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177699.4 | NP_808367.2 | 2 | 413 | |
GenBank mRNA | AB041045.1 | - | 2 | 307 | |
AK030737.1 | - | 2 | 375 | ||
BB858085.1 | - | 2 | 411 | ||
BC017144.1 | - | 2 | 352 | ||
BC060654.1 | - | 2 | 396 | 30354865 |