Gene Transcripts

Gene Symbol
Skint7
Entrez Gene ID
328505
Gene Name
selection and upkeep of intraepithelial T cells 7
Gene Aliases
C130057D23Rik
Chromosome Location
Chr. 4: 111972924 - 111988260 on Build GRCm38
UniGene ID
Mm.226771
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142775.1 NP_001136247.1 3 286
NM_177818.3 NP_808486.2 3 286
GenBank mRNA AK048392.1 - 3 181
AK132639.1 - 3 291
BB614300.1 - 3 286
EF494899.1 - 124
EF494900.1 - 124
EF494901.1 - 124
EF494902.1 - 124
EU099306.1 - 124

Gene Symbol
Skint8
Entrez Gene ID
639774
Gene Name
selection and upkeep of intraepithelial T cells 8
Gene Aliases
OTTMUSG00000009475
Chromosome Location
Chr. 4: 111919446 - 111950361 on Build GRCm38
UniGene ID
Mm.248185
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA EF494904.1 - 3 134

View Full Product Details