Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177909.5 | NP_808577.3 | 7 | 958 | |
GenBank mRNA | AK037058.1 | - | 7 | 942 | |
AK156494.1 | - | 7 | 919 | ||
AK172687.1 | - | 7 | 930 | ||
BC099942.1 | - | 7 | 932 | 30103308 | |
BC153825.1 | - | 7 | 936 | 40093280 |