Gene Transcripts

Gene Symbol
Ncapd3
Entrez Gene ID
78658
Gene Name
non-SMC condensin II complex, subunit D3
Gene Aliases
2810487N22Rik, 4632407J06Rik, AI195468, AU018739, B130055D15Rik, mKIAA0056
Chromosome Location
Chr. 9: 27030116 - 27096522 on Build GRCm38
UniGene ID
Mm.251777
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178113.3 NP_835214.2 5 966
GenBank mRNA AK014555.1 - 4 755
AK129046.1 - 4 492
AK139081.1 - 4 524
BC048190.1 - 5 966 6831876
DQ340802.1 - 4 621

View Full Product Details