Gene Transcripts

Gene Symbol
Tcfl5
Entrez Gene ID
277353
Gene Name
transcription factor-like 5 (basic helix-loop-helix)
Gene Aliases
AV260129, Figlb
Chromosome Location
Chr. 2: 180621956 - 180642691 on Build GRCm38
UniGene ID
Mm.473700
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178254.3 NP_839985.2 3 953
GenBank mRNA AK132721.1 - 3 610
AY234363.1 - 3 744
BC108399.1 - 3 953 6394526

View Full Product Details