Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GH9B5
Entrez Gene ID
838583
Gene Name
glycosyl hydrolase 9B5
Gene Aliases
AT1G19940, AtGH9B5, F6F9.1, F6F9_1, glycosyl hydrolase 9B5
Chromosome Location
Chr.1: 6918159 - 6921979 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332419.1 NP_001321680.1 6 - 7 1745 112
NM_101849.2 NP_173423.1 5 - 6 1426 112
GenBank mRNA AY048245.1 - 5 - 6 1322 112
AY113063.1 - 5 - 6 1213 112

View Full Product Details