Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AAE1
Entrez Gene ID
838644
Gene Name
acyl activating enzyme 1
Gene Aliases
AT1G20560, F5M15.12, F5M15_12, acyl activating enzyme 1
Chromosome Location
Chr.1: 7119671 - 7121871 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101906.4 NP_564116.1 1 - 2 346 59

View Full Product Details