Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT2G03290
Entrez Gene ID
814858
Gene Name
emp24/gp25L/p24 family/GOLD family protein
Gene Aliases
AT2G03290, T4M8.28, T4M8_28
Chromosome Location
Chr.2: 999406 - 1000688 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126380.5 NP_178428.3 3 - 4 514 154

View Full Product Details