Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUP4
Entrez Gene ID
839967
Gene Name
purine permease 4
Gene Aliases
AT1G30840, ATPUP4, T17H7.15, T17H7_15, purine permease 4
Chromosome Location
Chr.1: 10974383 - 10976298 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102821.3 NP_564365.1 1 - 1 1600 111

View Full Product Details