Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP97A3
Entrez Gene ID
840067
Gene Name
cytochrome P450, family 97, subfamily A, polypeptide 3
Gene Aliases
''cytochrome P450, AT1G31800, F5M6.19, F5M6_19, LUT5, LUTEIN DEFICIENT 5, cytochrome P450, family 97, polypeptide 3, polypeptide 3'', subfamily A
Chromosome Location
Chr.1: 11396402 - 11399714 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102914.3 NP_564384.1 5 - 6 858 120
GenBank mRNA AY056446.1 - 5 - 6 853 120
AY058173.1 - 5 - 6 858 120
AY142017.1 - 5 - 6 820 120

View Full Product Details