Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP97A3
Entrez Gene ID
840067
Gene Name
cytochrome P450, family 97, subfamily A, polypeptide 3
Gene Aliases
''cytochrome P450, AT1G31800, F5M6.19, F5M6_19, LUT5, LUTEIN DEFICIENT 5, cytochrome P450, family 97, polypeptide 3, polypeptide 3'', subfamily A
Chromosome Location
Chr.1: 11396402 - 11399714 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102914.3 NP_564384.1 8 - 9 1222 74
GenBank mRNA AY056446.1 - 8 - 9 1217 74
AY058173.1 - 8 - 9 1222 74
AY142017.1 - 8 - 9 1184 74

View Full Product Details