Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS5A
Entrez Gene ID
820367
Gene Name
ribosomal protein 5A
Gene Aliases
AML1, ARABIDOPSIS MINUTE-LIKE 1, AT3G11940, ATRPS5A, MEC18.11, ribosomal protein 5A
Chromosome Location
Chr.3: 3777824 - 3779545 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112027.2 NP_187800.1 1 - 2 203 74
NM_180233.3 NP_850564.1 2 - 3 196 74
GenBank mRNA AY045846.1 - 2 - 3 154 74
AY086938.1 - 1 - 2 203 74
AY091376.1 - 1 - 2 90 74

View Full Product Details